Sodium hydroxide 4.00 N in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Caustic soda

R7460000-4A R7460000-1A
RC7460-1CS 929.04 CAD
RC7460-1 RC7460-32
Sodium hydroxide 4.00 N in aqueous solution analytical reagent
Sodium hydroxide
Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 4.000 ± 0.001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR