Sodium hydroxide 2.00 N

Supplier: Ricca Chemical
Danger

R7455000-4A R7455000-1A R7455000-10F R7455000-500A
RC7455-1EA 255.96 CAD
RC7455-1 RC745532 RCR7455000-10F RCR7455000500A
Sodium hydroxide 2.00 N
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: ∼ 0°C
Density: 1.08
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 1.999-2.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR