Sodium hydroxide 10.0 N

Supplier: Ricca Chemical
Danger

R7470000-4A R7470000-20F R7470000-10F R7470000-500A
RC747001EA 339.7 CAD
RC747001 RC7470-5 RCR7470000-10F RCR7470000500A
Sodium hydroxide 10.0 N
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: ∼ 0°C
Density: 1.33
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 9.95-10.05 N at 20°C
Heavy Metals (as Pb) max 10 ppm
Iron (Fe) max 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR