Acetic acid (1 + 4) 20.0% (v/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R0138000-500A
R0138000-1A
R0138000-20F
RC138-16EA
77.42
CAD
RC138-16
RC138-32
RC138-5
Acetic acid (1 + 4) 20.0% (v/v) in aqueous solution
Acetic acid
Formula:
H₃CCOOH Boiling Pt: 196 °C (1013 hPa) Melting Pt: 54…56 °C Density: ≤1.07 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-927-2 UN: 2789 ADR: 8,II |
|
Specification Test Results
Appearance | Clear, colorless, acetic odor |
Assay at 20 °C (traceable to NIST SRM 84) | 20.0 ± 0.1 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...