Nitric acid 0.100 N (N/10)

Supplier: Ricca Chemical
Warning

R5400000-1A R5400000-2.5D
RC540032EA 97.96 CAD
RC540032 RC5400-5PT
Nitric acid 0.100 N (N/10)
Nitric acid
Formula: HNO₃
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR