Sodium hydroxide 0.125 N (N/8) in aqueous solution
Supplier: Ricca Chemical
R7356000-4A
R7356000-20F
RC73561EA
211.72
CAD
RC73561
RC73565
Sodium hydroxide 0.125 N (N/8) in aqueous solution
Sodium hydroxide
Formula:
NaOH Density: 1 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.1250 ± 0.0001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...