Acetic acid 25% (v/v) in aqueous solution (1 + 3)
Supplier: Ricca Chemical
|
Warning
|
R0138250-20F
R0138250-4A
R0138250-20E1
R0138250-500A
R0138250-1A
RC138.25-5EA
619.36
CAD
RC138.25-5
RC138.25-1
RC138.25-5HP
RC138.25-16
RC138.25-32
Acetic acid 25% (v/v) in aqueous solution (1 + 3)
Acetic acid
Formula:
H₃CCOOH Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2790 ADR: 8,III |
|
Specification Test Results
Appearance | Clear, colorless, acetic odor |
Assay at 20 °C (traceable to NIST SRM 84) | 25.00 ± 0.06 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...