Sulfuric acid 3.00 N in aqueous solution
Supplier: Ricca Chemical
|
Warning
|
R8320000-4A
R8320000-20F
RC8320-1EA
248.06
CAD
RC8320-1
RC8320-5
Sulfuric acid 3.00 N in aqueous solution
Sulfuric acid
Formula:
H₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 2796 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 3.000 ± 0.001 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...