Sodium hydroxide 3.00 N

Supplier: Ricca Chemical
Danger

R7459000-20F R7459000-10F R7459000-500A
RC7459-5EA 481.9 CAD
RC7459-5 RCR7459000-10F RCR7459000500A
Sodium hydroxide 3.00 N
Sodium hydroxide
Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: ∼ 0°C
Density: 1.12
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 2.999-3.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR