Sodium hydroxide 2.50 N analytical reagent
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Caustic soda
R7457000-4A
R7457000-1A
R7457000-20F
R7457000-10F
R7457000-500A
RC7457-1EA
219.62
CAD
RC7457-1
RC745732
RC7457-5
RCR7457000-10F
RCR7457000500A
Sodium hydroxide 2.50 N analytical reagent
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Boiling Pt: 1390 Melting Pt: 323 Density: 1.1 Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8 II Merck Index: 14,08627 |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Phenolphthalein) | 2.499-2.501 N at 20°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...