Ammonia solution 5.00 N analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

R0644000-1A R0644000-500A
RC64432EA 210.14 CAD
RC64432 RC644-16
Ammonia solution 5.00 N analytical reagent
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear, colorless, ammonia odor
Assay at 20 °C (traceable to NIST SRM 723) 5.005 ± 0.010 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR