Methanol 50% (v/v) in aqueous solution (1 + 1) analytical reagent

Supplier: Ricca Chemical
Danger

R4820000-4A R4820000-20F
RC48201EA 278.08 CAD
RC48201 RC4820-5
Methanol 50% (v/v) in aqueous solution (1 + 1) analytical reagent
Methanol
Formula: CH₃OH
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Assay 50 ± 1 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR