Sodium hypochlorite 2.5% (w/w) in aqueous solution

Supplier: Ricca Chemical

R7495100-500A R7495100-1A
RC7495.1-16EA 143.78 CAD
RC7495.1-16 RC7495.1-32
Sodium hypochlorite 2.5% (w/w) in aqueous solution
Sodium hypochlorite
Formula: ClNaO
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3

Order Now

Specification Test Results

Appearance Clear, colorless to greenish yellow, chlorine odor
Assay at 20 °C (traceable to NIST SRM 136) 2.55 ± 0.05 % (w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR