Iron(III) chloride 10% (w/v) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Iron trichloride, Ferric chloride

R3120000-500A R3120000-1A R3120000-120A
RC312016EA 116.92 CAD
RC312016 RC312032 RC31204
Iron(III) chloride 10% (w/v) in aqueous solution analytical reagent
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now

Specification Test Results

Appearance Clear, brown, no precipitate
Assay at 20 °C (traceable to NIST SRM 136) 10.00 ± 0.05 % (w/v)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR