Seawater (synthetic) ASTM D1141 heavy metal-free

Supplier: Ricca Chemical

R8363000-4A R8363000-20F
RC8363-1EA 278.08 CAD
RC8363-1 RC8363-5
Seawater (synthetic) ASTM D1141 heavy metal-free
Seawater (synthetic)

Substitute Ocean Water ASTM (D 1141) without heavy metals.
Synthetic Sea Water ASTM (D 665)
Substitute Ocean Water EPA (350.3)
Substitute Ocean Water EPA (353.2)
Substitute Ocean Water EPA (350.1)


Contains:
Water 7732-18-5 ACS/ASTM/USP/EP
Sodium Chloride 7647-14-5 ACS
Calcium Chloride Dihydrate 10035-04-8 ACS
Potassium Chloride 7447-40-7 ACS
Sodium Sulfate Anhydrous 7757-82-6 ACS
Magnesium Chloride Hexahydrate 7791-18-6 ACS
Strontium Chloride Hexahydrate 10025-70-4 ACS
Boric Acid 10043-35-3 ACS
Sodium Hydroxide 1310-73-2 Reagent
Sodium Bicarbonate 144-55-8 ACS
Sodium Fluoride 7681-49-4 ACS
Potassium Bromide 7758-02-3 ACS

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 8.225 ± 0.025 pH at 25.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR