Giemsa's stain stock solution

Supplier: Ricca Chemical
Danger

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

R3250000-500A R3250000-120A
RC325016EA 322.07 CAD
RC325016 RC32504
Giemsa's stain stock solution
Giemsa's stain

Giemsa Stain, Stock, for Blood and Tissue Staining, and Staining of Malarial Parasites in Blood.

Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081642
CAS Number: 51811-82-6
EINECS: 257-438-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Clear, dark violet, alcohol odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR