Fuchsin solution, decolorised R1
Supplier: Ricca Chemical
R3221000-500C
R3221000-1C
RC3221-16EA
238.58
CAD
RC3221-16
RC3221-32
Fuchsin solution, decolorised R1
Fuchsin solution, decolourised
UN:
1789 ADR: 8,III |
Specification Test Results
Absorbance (traceable to NIST SRM 930) | Conforms to control |
Absorabnce at 545 nm | Passes Test |
Appearance | Clear, colorless to slight yellow, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...