Benedict's reagent for qualitative determination of reducing sugars

Supplier: Ricca Chemical

R0930000-4A R0930000-500A R0930000-1A R0930000-120A
RC9301EA 336.54 CAD
RC9301 RC93016 RC93032 RC930-4
Benedict's reagent for qualitative determination of reducing sugars
Benedict's reagent
Storage Temperature: Ambient
MDL Number: MFCD00146068
CAS Number: 63126-89-6
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear, blue, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR