Hydrochloric acid 2.00 N

Supplier: Ricca Chemical
Danger

R3710000-4A R3710000-1A R3710000-20F R3710000-500A R3710000-10F
RC3710-1EA 312.84 CAD
RC3710-1 RC3710-32 RC3710-5 RC3710-16 RCR3710000-10F
Hydrochloric acid 2.00 N
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.999-2.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR