Phenol red 0.02% (w/v) in aqueous solution indicator

Supplier: Ricca Chemical

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

R5700000-500A R5700000-120A
RC570016EA 72.74 CAD
RC570016 RC57004
Phenol red 0.02% (w/v) in aqueous solution indicator
Phenol red
Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7

Order Now

Specification Test Results

Appearance clear, red, odorless
pH at 25 °C (traceable to NIST SRM 186 & 191) 7.20 ± 0.05 pH at 25.0 °C
Suitability for Use Yellow (pH 8.2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR