Orthoboric acid 2% (w/v) in aqueous solution

Supplier: Ricca Chemical
Danger

R1065000-4A R1065000-1A R1065000-20F R1065000-500A
RC10651EA 260.7 CAD
RC10651 RC106532 RCR1065000-20F RCR1065000500A
Orthoboric acid 2% (w/v) in aqueous solution
ortho-Boric acid
Formula: B(OH)₃
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.94-2.06 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR