Hydrochloric acid 0.500 N (N/2) in 2-propanol

Supplier: Ricca Chemical
Danger

R3805000-4A R3805000-1A
RC38051EA 429.76 CAD
RC38051 RC380532
Hydrochloric acid 0.500 N (N/2) in 2-propanol
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless, HCl/IPA odor
Assay at 20 °C (traceable to NIST SRM 84) 0.5000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR