Acetic acid 5% (v/v) in aqueous solution (1 + 19)

Supplier: Ricca Chemical
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

R0130000-4A R0130000-500A R0130000-1A R0130000-10F R0130000-20F
RC1301EA 211.72 CAD
RC1301 RC13016 RC13032 RCR0130000-10F RCR0130000-20F
Acetic acid 5% (v/v) in aqueous solution (1 + 19)
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Thymol Blue) 4.94-5.06 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR