Acetic acid 2% (v/v) in aqueous solution (1 + 49) analytical reagent

Supplier: Ricca Chemical

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

R0110000-500A R0110000-1A R0110000-4A
RC11016EA 61.62 CAD
RC11016 RC11032 RC110-1
Acetic acid 2% (v/v) in aqueous solution (1 + 49) analytical reagent
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, slight acetic odor
Assay at 20 °C (traceable to NIST SRM 84) 2.00 ± 0.06 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR