Acetic acid 1% v/v aqueous solution (1 + 99) analytical reagent

Supplier: Ricca Chemical

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

R0100000-500A R0100000-1A R0100000-4A
RC10016EA 61.62 CAD
RC10016 RC10032 RC100-1
Acetic acid 1% v/v aqueous solution (1 + 99) analytical reagent
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118
Melting Pt: 17
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, slight acetic odor
Assay at 20 °C (traceable to NIST SRM 84) 1.00 ± 0.06 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR