Ammonia solution 0.500 N (N/2)

Supplier: Ricca Chemical

R0641000-1A R0641000-4A R0641000-500A
RC64132EA 180.12 CAD
RC64132 RC641-1 RCR0641000500A
Ammonia solution 0.500 N (N/2)
Ammonia solution
Formula: NH₃
Melting Pt: 0.0°C
Density: 1
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Methyl Red) 0.4997-0.5007 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR