Nitric acid 10% (v/v) in aqueous solution (1 + 9)
Supplier: Ricca Chemical
|
Danger
|
R5320000-1A
R5320000-20F
R5320000-4A
R5320000-2.5D
R5320000-20E1
RC5320-32EA
180.12
CAD
RC5320-32
RCR5320000-20F
RCR5320000-4A
RCR53200002.5D
RCR532000020E1
Nitric acid 10% (v/v) in aqueous solution (1 + 9)
Nitric acid
Formula:
HNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 3264 ADR: 8,III |
Specification Test Results
Appearance | Colorless to light yellow liquid |
Assay (vs. Sodium Hydroxide/Phenolphthalein) | 9.9-10.1 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...