Hydrochloric acid 0.0200 N (N/50)

Supplier: Ricca Chemical

R3595000-20F R3595000-4A R3595000-1A
RC3595-5EA 455.04 CAD
RC3595-5 RC35951 RC359532
Hydrochloric acid 0.0200 N (N/50)
Hydrochloric acid
Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.02000 ± 0.00005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR