Sodium carbonate 10% (w/v) in aqueous solution

Supplier: Ricca Chemical

R7160000-4A R7160000-500A R7160000-1A
RC71601EA 222.78 CAD
RC71601 RC716016 RC716032
Sodium carbonate 10% (w/v) in aqueous solution
Sodium carbonate
Formula: Na₂CO₃
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 10.0 ± 0.3 % (w/v)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR