Uranium AA Standard, 1 ml = 1 mg U (1000 ppm U)UO3 in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RAU1KN00-100N RAU1KN00-500N
RCAU1KN-500EA 1545.05 CAD
RCAU1KN-100 RCAU1KN-500
Uranium AA Standard, 1 ml = 1 mg U (1000 ppm U)UO3 in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Uranium Standards