Hydrochloric acid 0.0100 N (N/100) analytical reagent

Supplier: Ricca Chemical

Synonyms: Hydrochloric acid solution, Muriatic acid

R3590000-20F R3590000-4A R3590000-1A R3590000-120A R3590000-10F R3590000-500A
RC3590-5EA 587.76 CAD
RC3590-5 RC35901 RC359032 RC3590-4 RCR3590000-10F RCR3590000500A
Hydrochloric acid 0.0100 N (N/100) analytical reagent
Hydrochloric acid
Formula: HCl
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 0.00995-0.01005 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR