Thallium ICP Standard, 1 ml = 10 mg Tl (10000 ppm Tl)Tl in 5% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPTL10KN-100N RPTL10KN-50N RPTL10KN-500N
RCPTL10KN-100EA 382.36 CAD
RCPTL10KN-100 RCPTL10KN-50 RCPTL10KN-500
Thallium ICP Standard, 1 ml = 10 mg Tl (10000 ppm Tl)Tl in 5% HNO₃, Ricca Chemical Company
Standards Cationic Standards Terbium Standards