Potassium chloride 3 M aqueous solution Electrodes_filling

Supplier: Ricca Chemical

Synonyms: Potassiumchloride

R5899000-500A R5899000-120A
RC589916EA 121.66 CAD
RC589916 RC58994
Potassium chloride 3 M aqueous solution Electrodes_filling
Potassium chloride

Filling solution for ROSS pH combination and reference electrodes.

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Molarity 3.00 ± 0.07 M at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR