Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, Reagent Grade

Supplier: Ricca Chemical

R3819000-4B R3819000-500B R3819000-1B
RC3819-1EA 382.36 CAD
RC3819-1 RC3819-16 RC3819-32
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, Reagent Grade
Hydrogen peroxide
Formula: H₂O₂
Boiling Pt: 100 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40) 3.3 ± 0.1 % (w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR