Sulfuric acid 20% (w/w) in aqueous solution

Supplier: Ricca Chemical
Danger

R8166000-500C1 R8166000-2.5D R8166000-20F R8166000-4C1
RC8166-16EA 71.1 CAD
RC8166-16 RC8166-5PT RCR8166000-20F RCR8166000-4C1
Sulfuric acid 20% (w/w) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 19.95-20.05 % (w/w)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR