Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for quantitative determination of reducing sugars

Supplier: Ricca Chemical
Danger

R3000000-4A R3000000-20F R3000000-500A R3000000-1A
RC3000-1EA 425.02 CAD
RC3000-1 RC3000-5 RC300016 RC300032
Fehling's reagent II (L(+)-potassium sodium tartrate in sodium hydroxide solution) for quantitative determination of reducing sugars
Fehling's reagent II (L(+)-Potassium sodium tartrate in sodium hydroxide solution)
Density: 1.2 g/cm³ (20 °C)
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR