Sulfuric acid 20% (v/v) in aqueous solution (1 + 4), Reagent Grade

Supplier: Ricca Chemical
Danger

R8165000-500A R8165000-1A R8165000-4A R8165000-20F
RC8165-16EA 77.42 CAD
RC8165-16 RC8165-32 RC8165-1 RC8165-5
Sulfuric acid 20% (v/v) in aqueous solution (1 + 4), Reagent Grade
Sulfuric acid
Formula: H₂SO₄
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 20.00 ± 0.03 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR