Sulfuric acid 0.250 N (N/4) in aqueous solution

Supplier: Ricca Chemical

R8270000-4A R8270000-20F R8270000-1A
RC82701EA 191.18 CAD
RC82701 RC82705 RC8270-32
Sulfuric acid 0.250 N (N/4) in aqueous solution
Sulfuric acid
Formula: H₂SO₄
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.2500 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR