Sulfuric acid 10% (v/v) in aqueous solution (1 + 9)

Supplier: Ricca Chemical
Warning

R8150000-500A R8150000-1A R8150000-4A
RC815016EA 91.64 CAD
RC815016 RC8150-32 RC8150-1
Sulfuric acid 10% (v/v) in aqueous solution (1 + 9)
Sulfuric acid
Formula: H₂SO₄
Boiling Pt: 101 °C (1013 hPa)
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II
Merck Index: 13,09064

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 10.00 ± 0.03 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR