Sodium hydroxide 50% (w/v) in aqueous solution analytical reagent, for Orsat gas analysis

Supplier: Ricca Chemical
Danger

R7283000-20F R7283000-10F R7283000-1A R7283000-500A
RC7283-5EA 654.12 CAD
RC7283-5 RC7283-2.5 RCR7283000-1A RCR7283000500A
Sodium hydroxide 50% (w/v) in aqueous solution analytical reagent, for Orsat gas analysis
Sodium hydroxide
Formula: NaOH
Boiling Pt: ~100 °C
Melting Pt: ~0 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sulfuric Acid/Phenolphthalein) 49.92 - 50.08 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR