Lithium ICP Standard, 1 mL = 1 mg Li (1000 ppm Li)Li₂CO₃ in 3% HNO₃, Ricca Chemical Company
Supplier: RICCA Chemical Company
RPLI1KN0-500N RPLI1KN0-100N
RCPLI1KN-500EA 460.44 CAD
RCPLI1KN-100 RCPLI1KN-500
Lithium ICP Standard, 1 mL = 1 mg Li (1000 ppm Li)Li₂CO₃ in 3% HNO₃, Ricca Chemical Company
Standards Cationic Standards Lithium Standards