Sodium hydroxide 30% (w/v) in aqueous solution
Supplier: Ricca Chemical
|
Danger
|
R7274000-4A
R7274000-1A
RC7274-1EA
300.2
CAD
RC7274-1
RC7274-32
Sodium hydroxide 30% (w/v) in aqueous solution
Sodium hydroxide
Formula:
NaOH Boiling Pt: 119…135 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 7.50 ± 0.05 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...