Sodium hydroxide 25% (w/v) in aqueous solution (250 g/L)

Supplier: Ricca Chemical
Danger

R7273000-500A R7273000-1A R7273000-20F
RC727316EA 101.12 CAD
RC727316 RC7273-32 RC7273-5
Sodium hydroxide 25% (w/v) in aqueous solution (250 g/L)
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 25.00 ± 0.06 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR