Color Standard, USP / EP Yellow Primary Solution, Ferric Chloride Solution, Ricca Chemical Company
Supplier: Ricca Chemical
R3110000-1C R3110000-500C R3110000-120C
RC311032EA 513.5 CAD
RC311032 RC311016 RC3110-4
Color Standard, USP / EP Yellow Primary Solution, Ferric Chloride Solution, Ricca Chemical Company
Standards Colour Reference and Spectroscopy Standards
1 mL = 45.0 mg FeCl₃.6H₂O