Fehling's reagent I (copper(II) sulfate solution) for quantitative determination of reducing sugars

Supplier: Ricca Chemical

Synonyms: Fehling A solution, Cupric sulfate pentahydrate, Copper (II) sulfate 5-hydrate

R3010000-4A R3010000-20F R3010000-500A R3010000-1A
RC3010-1EA 216.46 CAD
RC3010-1 RC3010-5 RC301016 RC301032
Fehling's reagent I (copper(II) sulfate solution) for quantitative determination of reducing sugars
Fehling's reagent I (Copper (II) sulphate solution)
Formula: CuSO₄
MW: 159.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9 III

Order Now

Specification Test Results

Appearance Clear, blue odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.2775 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR