Sodium hypochlorite 5% available chlorine in aqueous solution analytical reagent

Supplier: Ricca Chemical
Danger

R7495500-4B R7495500-1B R7495500-500B
RC7495.5-1EA 426.6 CAD
RC7495.5-1 RC7495.5-32 89423-884
Sodium hypochlorite 5% available chlorine in aqueous solution analytical reagent
Sodium hypochlorite
Formula: NaOCl
MW: 74.44 g/mol
Melting Pt: -6
Density: 1.22…1.25 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00011120
CAS Number: 7681-52-9
EINECS: 231-668-3
UN: 1791
ADR: 8 II
Merck Index: 13,08702

Order Now

Specification Test Results

Appearance Clear, colorless to slight yellow, strong bleach odor
Assay at 20 °C (traceable to NIST SRM 136) 5.00 ± 0.25 % (w/w) available Chlorine


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR