Hydrochloric acid 3.00 N analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

R3720000-1A R3720000-20F R3720000-4A
RC3720-32EA 105.86 CAD
RC3720-32 RC3720-5 RC3720-1
Hydrochloric acid 3.00 N analytical reagent
Hydrochloric acid
Formula: HCl
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, slight Hydrochloric odor
Assay at 20 °C (traceable to NIST SRM 84) 3.000 ± 0.001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR