Acetic acid 50% (v/v) (1 + 1)

Supplier: Ricca Chemical
Danger

R0140000-4C R0140000-500C R0140000-4F R0140000-1C
RC140-1EA 410.8 CAD
RC140-1 RC140-16 RC140-1CT RC140-32
Acetic acid 50% (v/v) (1 + 1)
Acetic acid
Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, strong Acetic odor
Assay at 20 °C (traceable to NIST SRM 84) 50.00 ± 0.05 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR