Nitric acid 50% (v/v) in aqueous solution (1 + 1)
Supplier: Ricca Chemical
|
Danger
|
R5350000-500C1
R5350000-1C1
RC535016EA
167.48
CAD
RC535016
RC535032
Nitric acid 50% (v/v) in aqueous solution (1 + 1)
Nitric acid
Formula:
HNO₃ Boiling Pt: ∼86 °C (1013 hPa) Melting Pt: –42 °C Density: 1.2…1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, Nitric odor |
Assay at 20 °C (traceable to NIST SRM 84) | 50.0 ± 0.3 % (v/v) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...