Nitric acid 20% (v/v) in aqueous solution (1 + 4)

Supplier: Ricca Chemical
Danger

R5326000-500C R5326000-2.5D
RC5326-16EA 148.52 CAD
RC5326-16 RCR53260002.5D
Nitric acid 20% (v/v) in aqueous solution (1 + 4)
Nitric acid
Formula: HNO₃
Boiling Pt: ~ 86 °C (1013 hPa)
Melting Pt: -42 °C
Density: 1.2 to 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 19.9-20.1 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR